Assay Details
| Gene Symbol: LOC100793233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254093.2 | NP_001241022.1 | 3 - 4 | 147 | 106 | |
| GenBank mRNA | BT092970.1 | - | 3 - 4 | 149 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT092970.1, NM_001254093.2 transcripts.
Set Membership: |
|

LOC100793233
Genomic Map