Assay Details
| Gene Symbol: LOC100803227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254145.2 | NP_001241074.1 | 2 - 3 | 717 | 70 | |
| GenBank mRNA | BT097739.1 | - | 2 - 3 | 719 | 70 | |
More Information
Set Membership: |
|

LOC100803227
Genomic Map