Assay Details
| Gene Symbol: LOC100802974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254163.2 | NP_001241092.1 | 1 - 2 | 64 | 82 | |
| GenBank mRNA | BT094039.1 | - | 1 - 2 | 66 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT094039.1, NM_001254163.2 transcripts.
Set Membership: |
|

LOC100802974
Genomic Map