Assay Details
| Gene Symbol: LOC100785827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254196.2 | NP_001241125.1 | 4 - 5 | 677 | 70 | |
| GenBank mRNA | BT093942.1 | - | 4 - 5 | 678 | 70 | |
| Gene Symbol: LOC100780029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287177.1 | - | 4 - 5 | 702 | 70 | |
More Information
Set Membership: |
|

LOC100785827
Genomic Map