Assay Details
| Gene Symbol: PHT1-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254198.1 | NP_001241127.1 | 1 | 223 | 66 | |
| GenBank mRNA | FJ797412.1 | - | 1 | 223 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHT1-12| Gene Symbol: PHT1-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254198.1 | NP_001241127.1 | 1 | 223 | 66 | |
| GenBank mRNA | FJ797412.1 | - | 1 | 223 | 66 | |
Set Membership: |
|