Assay Details
| Gene Symbol: LOC100813490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254278.2 | NP_001241207.1 | 10 - 11 | 1020 | 87 | |
| GenBank mRNA | BT095427.1 | - | 10 - 11 | 1022 | 87 | |
More Information
Set Membership: |
|

LOC100813490
Genomic Map