Assay Details
| Gene Symbol: LOC100813490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254278.2 | NP_001241207.1 | 1 - 2 | 89 | 134 | |
| GenBank mRNA | BT095427.1 | - | 1 - 2 | 91 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT095427.1, NM_001254278.2 transcripts.
Set Membership: |
|

LOC100813490
Genomic Map