Assay Details
| Gene Symbol: LOC100813490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254278.2 | NP_001241207.1 | 4 - 5 | 285 | 93 | |
| GenBank mRNA | BT095427.1 | - | 4 - 5 | 287 | 93 | |
More Information
Set Membership: |
|

LOC100813490
Genomic Map