Assay Details
| Gene Symbol: GMFT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254292.1 | NP_001241221.1 | 3 - 4 | 293 | 169 | |
| GenBank mRNA | AB550121.1 | - | 3 - 4 | 293 | 169 | |
| FJ573244.1 | - | 3 - 4 | 293 | 169 | ||
More Information
Set Membership: |
|

GMFT1B
Genomic Map