Assay Details
| Gene Symbol: LOC100784566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254300.2 | NP_001241229.1 | 9 - 10 | 945 | 102 | |
| GenBank mRNA | BT093996.1 | - | 9 - 10 | 947 | 102 | |
More Information
Set Membership: |
|

LOC100784566
Genomic Map