Assay Details
| Gene Symbol: LOC100789704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254411.1 | NP_001241340.1 | 1 - 2 | 2515 | 61 | |
| GenBank mRNA | FJ014785.1 | - | 1 - 2 | 2515 | 61 | |
More Information
Set Membership: |
|

LOC100789704
Genomic Map