Assay Details
| Gene Symbol: PHT1-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254471.1 | NP_001241400.1 | 1 | 222 | 64 | |
| GenBank mRNA | FJ797413.1 | - | 1 | 222 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHT1-13| Gene Symbol: PHT1-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254471.1 | NP_001241400.1 | 1 | 222 | 64 | |
| GenBank mRNA | FJ797413.1 | - | 1 | 222 | 64 | |
Set Membership: |
|