Assay Details
| Gene Symbol: LOC100814321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254631.1 | NP_001241560.1 | 2 - 3 | 82 | 80 | |
| GenBank mRNA | BT099442.1 | - | 10 - 11 | 1336 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001254631.1 transcripts.
Set Membership: |
|

LOC100814321
Genomic Map