Assay Details
| Gene Symbol: LOC100814321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254631.1 | NP_001241560.1 | 6 - 7 | 362 | 111 | |
| GenBank mRNA | BT099442.1 | - | 6 - 7 | 1056 | 111 | |
More Information
Set Membership: |
|

LOC100814321
Genomic Map