Assay Details
| Gene Symbol: LOC100805114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254679.2 | NP_001241608.1 | 2 - 3 | 821 | 101 | |
| GenBank mRNA | BT097267.1 | - | 2 - 3 | 823 | 101 | |
More Information
Set Membership: |
|

LOC100805114
Genomic Map