Assay Details
| Gene Symbol: LOC100805114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254679.2 | NP_001241608.1 | 3 - 4 | 896 | 122 | |
| GenBank mRNA | BT097267.1 | - | 3 - 4 | 898 | 122 | |
More Information
Set Membership: |
|

LOC100805114
Genomic Map