Assay Details
| Gene Symbol: LOC100779031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255039.2 | NP_001241968.1 | 2 - 3 | 429 | 86 | |
| GenBank mRNA | BT096630.1 | - | 2 - 3 | 431 | 86 | |
More Information
Set Membership: |
|

LOC100779031
Genomic Map