Assay Details
| Gene Symbol: LOC100789278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255066.2 | NP_001241995.1 | 10 - 11 | 799 | 75 | |
| GenBank mRNA | AK285981.1 | - | 10 - 11 | 798 | 75 | |
| BT094236.1 | - | 10 - 11 | 801 | 75 | ||
More Information
Set Membership: |
|

LOC100789278
Genomic Map