Assay Details
| Gene Symbol: LOC100781599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255070.1 | NP_001241999.1 | 5 - 6 | 564 | 72 | |
| GenBank mRNA | BT095543.1 | - | 5 - 6 | 564 | 72 | |
More Information
Set Membership: |
|

LOC100781599
Genomic Map