Assay Details
| Gene Symbol: LOC100775215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255131.2 | NP_001242060.1 | 6 - 7 | 501 | 78 | |
| GenBank mRNA | BT093906.1 | - | 6 - 7 | 503 | 78 | |
More Information
Set Membership: |
|

LOC100775215
Genomic Map