Assay Details
| Gene Symbol: LOC100775215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255131.2 | NP_001242060.1 | 7 - 8 | 641 | 104 | |
| GenBank mRNA | BT093906.1 | - | 7 - 8 | 643 | 104 | |
More Information
Set Membership: |
|

LOC100775215
Genomic Map