Assay Details
| Gene Symbol: LOC100781400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255304.2 | NP_001242233.1 | 6 - 7 | 556 | 123 | |
| GenBank mRNA | BT098825.1 | - | 6 - 7 | 558 | 123 | |
| Gene Symbol: LOC100814621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245228.1 | - | 6 - 7 | 698 | 123 | |
More Information
Set Membership: |
|

LOC100781400
Genomic Map