Assay Details
| Gene Symbol: LOC100810842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255308.2 | NP_001242237.1 | 4 - 5 | 293 | 120 | |
| GenBank mRNA | BT096134.1 | - | 4 - 5 | 295 | 120 | |
More Information
Set Membership: |
|

LOC100810842
Genomic Map