Assay Details
| Gene Symbol: LOC100810338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255344.2 | NP_001242273.1 | 5 - 6 | 1013 | 112 | |
| GenBank mRNA | BT098195.1 | - | 5 - 6 | 1015 | 112 | |
More Information
Set Membership: |
|

LOC100810338
Genomic Map