Assay Details
| Gene Symbol: LOC100792444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255443.2 | NP_001242372.1 | 4 - 5 | 360 | 153 | |
| GenBank mRNA | BT097143.1 | - | 4 - 5 | 362 | 153 | |
More Information
Set Membership: |
|

LOC100792444
Genomic Map