Assay Details
| Gene Symbol: LOC100787120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255664.2 | NP_001242593.1 | 1 - 2 | 213 | 126 | |
| GenBank mRNA | BT097822.1 | - | 6 - 7 | 843 | 126 | |
| BT097827.1 | - | 1 - 2 | 215 | 126 | ||
More Information
Set Membership: |
|

LOC100787120
Genomic Map