Assay Details
| Gene Symbol: RSDV1-LIKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255676.2 | NP_001242605.1 | 4 - 5 | 494 | 102 | |
| GenBank mRNA | BT094519.1 | - | 4 - 5 | 496 | 102 | |
More Information
Set Membership: |
|

RSDV1-LIKE
Genomic Map