Assay Details
| Gene Symbol: LOC100807362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255731.2 | NP_001242660.1 | 2 - 3 | 737 | 75 | |
| GenBank mRNA | BT094341.1 | - | 2 - 3 | 739 | 75 | |
More Information
Set Membership: |
|

LOC100807362
Genomic Map