Assay Details
| Gene Symbol: LOC100814276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255744.1 | NP_001242673.1 | 1 - 2 | 135 | 61 | |
| GenBank mRNA | BT096571.1 | - | 1 - 2 | 135 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT096571.1, NM_001255744.1 transcripts.
Set Membership: |
|

LOC100814276
Genomic Map