Assay Details
| Gene Symbol: LOC100780764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255835.1 | NP_001242764.1 | 1 - 2 | 43 | 100 | |
| GenBank mRNA | BT095374.1 | - | 1 - 2 | 43 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT095374.1, NM_001255835.1 transcripts.
Set Membership: |
|

LOC100780764
Genomic Map