Assay Details
| Gene Symbol: LOC100808414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255839.1 | NP_001242768.1 | 4 - 5 | 446 | 98 | |
| GenBank mRNA | BT099034.1 | - | 4 - 5 | 446 | 98 | |
More Information
Set Membership: |
|

LOC100808414
Genomic Map