Assay Details
| Gene Symbol: LOC100805618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255911.1 | NP_001242840.1 | 13 - 14 | 1230 | 93 | |
| GenBank mRNA | BT097916.1 | - | 13 - 14 | 1230 | 93 | |
More Information
Set Membership: |
|

LOC100805618
Genomic Map