Assay Details
| Gene Symbol: PIN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280570.1 | NP_001267499.1 | 1 - 2 | 1213 | 107 | |
| GenBank mRNA | KC344371.1 | - | 1 - 2 | 1213 | 107 | |
More Information
Set Membership: |
|

PIN1B
Genomic Map