Assay Details
| Gene Symbol: PLDZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288593.1 | NP_001275522.1 | 14 - 15 | 2568 | 157 | |
| GenBank mRNA | KF542816.1 | - | 14 - 15 | 2568 | 157 | |
More Information
Set Membership: |
|

PLDZ
Genomic Map