Assay Details
| Gene Symbol: LOC100806689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289215.1 | NP_001276144.1 | 12 - 13 | 1596 | 90 | |
| GenBank mRNA | AK287294.1 | - | 12 - 13 | 1668 | 90 | |
| FJ594403.1 | - | 12 - 13 | 1596 | 90 | ||
More Information
Set Membership: |
|

LOC100806689
Genomic Map