Assay Details
| Gene Symbol: LOC100815664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289227.1 | NP_001276156.1 | 2 - 3 | 230 | 93 | |
| GenBank mRNA | AK244741.1 | - | 2 - 3 | 229 | 93 | |
| BT094317.1 | - | 2 - 3 | 230 | 93 | ||
More Information
Set Membership: |
|

LOC100815664
Genomic Map