Assay Details
| Gene Symbol: LOC100811134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289291.1 | NP_001276220.1 | 6 - 7 | 608 | 61 | |
| GenBank mRNA | AK286914.1 | - | 6 - 7 | 623 | 61 | |
| BT096585.1 | - | 6 - 7 | 608 | 61 | ||
More Information
Set Membership: |
|

LOC100811134
Genomic Map