Assay Details
| Gene Symbol: LOC100796900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289365.1 | NP_001276294.1 | 6 - 7 | 493 | 97 | |
| GenBank mRNA | BT098540.1 | - | 8 - 9 | 799 | 97 | |
More Information
Set Membership: |
|

LOC100796900
Genomic Map