Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244945.1 | - | 3 - 4 | 2343 | 100 | |
| AK245027.1 | - | 3 - 4 | 2343 | 100 | ||
| FJ968750.1 | - | 3 - 4 | 931 | 100 | ||
More Information
Set Membership: |
|

STS
Genomic Map