Assay Details
| Gene Symbol: LOC100786990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK285697.1 | - | 6 - 7 | 717 | 67 | |
| BT092170.1 | - | 6 - 7 | 723 | 67 | ||
| BT096780.1 | - | 6 - 7 | 725 | 67 | ||
| Gene Symbol: LOC100817464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK286688.1 | - | 6 - 7 | 739 | 67 | |
More Information
Set Membership: |
|

LOC100786990
Genomic Map