Assay Details
| Gene Symbol: G4DT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249061.1 | NP_001235990.1 | 11 - 12 | 1078 | 96 | |
| GenBank mRNA | AB434690.1 | - | 11 - 12 | 1078 | 96 | |
More Information
Set Membership: |
|

G4DT
Genomic Map