Assay Details
| Gene Symbol: G4DT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249061.1 | NP_001235990.1 | 3 - 4 | 375 | 77 | |
| GenBank mRNA | AB434690.1 | - | 3 - 4 | 375 | 77 | |
| BT092081.1 | - | 3 - 4 | 468 | 77 | ||
More Information
Set Membership: |
|

G4DT
Genomic Map