Assay Details
| Gene Symbol: LOC100796208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255138.2 | NP_001242067.1 | 4 - 5 | 659 | 133 | |
| GenBank mRNA | BT094436.1 | - | 4 - 5 | 661 | 133 | |
| Gene Symbol: LOC100787712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244224.1 | - | 4 - 5 | 690 | 133 | |
More Information
Set Membership: |
|

LOC100796208
Genomic Map