Assay Details
| Gene Symbol: LOC100785073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255668.1 | NP_001242597.1 | 1 - 2 | 125 | 105 | |
| GenBank mRNA | BT097712.1 | - | 1 - 2 | 125 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT097712.1, NM_001255668.1 transcripts.
Set Membership: |
|

LOC100785073
Genomic Map