Assay Details
| Gene Symbol: LOC100788665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255706.1 | NP_001242635.1 | 8 - 9 | 835 | 110 | |
| GenBank mRNA | BT097643.1 | - | 8 - 9 | 835 | 110 | |
More Information
Set Membership: |
|

LOC100788665
Genomic Map