Assay Details
| Gene Symbol: LOC100787304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253265.2 | NP_001240194.1 | 1 - 2 | 209 | 105 | |
| GenBank mRNA | AK286426.1 | - | 1 - 2 | 169 | 105 | |
| BT096395.1 | - | 1 - 2 | 211 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT096395.1, NM_001253265.2 transcripts.
Set Membership: |
|

LOC100787304
Genomic Map