Assay Details
| Gene Symbol: LOC100803483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254146.2 | NP_001241075.1 | 1 - 2 | 238 | 94 | |
| GenBank mRNA | BT093516.1 | - | 1 - 2 | 212 | 94 | |
| BT094106.1 | - | 1 - 2 | 240 | 94 | ||
More Information
Set Membership: |
|

LOC100803483
Genomic Map