Assay Details
| Gene Symbol: LOC100798499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255084.1 | NP_001242013.1 | 1 - 2 | 205 | 114 | |
| GenBank mRNA | BT095535.1 | - | 1 - 2 | 205 | 114 | |
More Information
Set Membership: |
|

LOC100798499
Genomic Map