Assay Details
| Gene Symbol: LOC100786504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255243.2 | NP_001242172.1 | 4 - 5 | 690 | 121 | |
| GenBank mRNA | BT097599.1 | - | 4 - 5 | 692 | 121 | |
More Information
Set Membership: |
|

LOC100786504
Genomic Map