Assay Details
| Gene Symbol: LOC100817010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255407.1 | NP_001242336.1 | 5 - 6 | 1005 | 88 | |
| GenBank mRNA | BT093690.1 | - | 5 - 6 | 1005 | 88 | |
More Information
Set Membership: |
|

LOC100817010
Genomic Map